accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66353-47-7

IUPAC Name: 1-(2,4-dichlorophenyl)butan-1-one
Synonyms: 1-(2,4-dichlorophenyl)butan-1-one
Molecular Weight (g/mol): 217.09
Molecular Formula: C10H10Cl2O
InChi Key: FXCJYMLJTUZGDU-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=C(Cl)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results