accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6636-78-8

IUPAC Name: 2-chloropyridin-3-ol
Synonyms: 3-pyridinol, 2-chloro-
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: RSOPTYAZDFSMTN-UHFFFAOYSA-N
SMILES: OC1=CC=CN=C1Cl
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)