CAS RN 66369-14-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Supelco™ Derivatized Silica Gel, 15-40 μm
Prepared by the reaction of silica gel with n-octadecyltrichlorosilane. After hydrolysis of the unreacted chloro groups, hydroxyl groups are blocked with trimethylsilyl groups.