accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66376-36-1

IUPAC Name: (4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid
Synonyms: alendronate alendronic acid monosodium salt trihydrate alendronate sodium aminohydroxybutane bisphosphonate acide alendronique alendronate sodium hydrate sodium, alendronate alendronate sodium - chemical alendronate monosodium salt, trihydrate aminohydroxybutylidene biphosphonate monosodium salt trihydrate
Molecular Weight (g/mol): 249.1
Molecular Formula: C4H13NO7P2
InChi Key: OGSPWJRAVKPPFI-UHFFFAOYSA-N
SMILES: NCCCC(O)(P(O)(O)=O)P(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results