accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3HNOH3C

CAS RN 6638-79-5

IUPAC Name: hydrogen methoxy(methyl)amine chloride
Synonyms: hydrogen N,O-dimethylhydroxylamine chloride
Molecular Weight (g/mol): 97.54
Molecular Formula: C2H8ClNO
InChi Key: USZLCYNVCCDPLQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNOC
Molecular Weight (g/mol) 
  • (10)
  • (4)
Quantity 
  • (1)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (7)
Physical Form 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (4)
  • (1)
  • (3)
  • (6)
  • (10)
  • (4)
  • (1)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (3)
  • (1)