accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 663949-34-6

H3CBr

CAS RN 663949-34-6

IUPAC Name: 1-bromo-3-octylbenzene
Synonyms: 1-bromo-3-octylbenzene
Molecular Weight (g/mol): 269.23
Molecular Formula: C14H21Br
InChi Key: SVXNIBGYTOIBPP-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC(Br)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results