CAS RN 6640-27-3
IUPAC Name:
2-chloro-4-methylphenol
Synonyms:
2-chloro-4-methylphenol C7H7ClO
Molecular Weight (g/mol):
142.58
Molecular Formula:
C7H7ClO
InChi Key:
AQJFATAFTQCRGC-UHFFFAOYSA-N
SMILES:
CC1=CC=C(O)C(Cl)=C1
1
–
1
of
1
results
1
2-Chloro-p-cresol 97.0+%, TCI America™
CAS: 6640-27-3 Molecular Formula: C7H7ClO Molecular Weight (g/mol): 142.582 InChI Key: AQJFATAFTQCRGC-UHFFFAOYSA-N Synonym: 2-Chloro-4-methylphenol PubChem CID: 14851 ChEBI: CHEBI:79732 IUPAC Name: 2-chloro-4-methylphenol SMILES: CC1=CC(=C(C=C1)O)Cl