accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6641-83-4

CH3H3COOHO

CAS RN 6641-83-4

IUPAC Name: 2-methyl-4-oxopentanoic acid
Synonyms: 2-methyl-4-oxopentanoic acid
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: UZTJTTKEYGHTNM-UHFFFAOYNA-N
SMILES: CC(CC(C)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results