accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OOOClN

CAS RN 66441-23-4

IUPAC Name: ethyl 2-{4-[(6-chloro-1,3-benzoxazol-2-yl)oxy]phenoxy}propanoate
Synonyms: depon epa pesticide chemical code 128701 furore caswell no. 431C fenoxaprop ethyl Whip acclaim fenoxaprop ethyl ester PUMA ethyl fenoxaprop
Molecular Weight (g/mol): 361.78
Molecular Formula: C18H16ClNO5
InChi Key: PQKBPHSEKWERTG-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)OC1=CC=C(OC2=NC3=CC=C(Cl)C=C3O2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results