CAS RN 66455-27-4
CAS RN 66455-27-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MP Biomedicals, Inc α-1-Acid Glycoprotein from Human Plasma, MP Biomedicals™
CAS: 66455-27-4 MDL Number: MFCD00131224 Synonym: a1-AG,orosomucoid,a1-seromucoid,AGP