accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66476-44-6

IUPAC Name: N-methyl-N-[(trifluoromethyl)sulfanyl]aniline
Synonyms: N-methyl-N-[(trifluoromethyl)sulfanyl]aniline
Molecular Weight (g/mol): 207.21
Molecular Formula: C8H8F3NS
InChi Key: TYFGNILTFVMYGX-UHFFFAOYSA-N
SMILES: CN(SC(F)(F)F)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results