accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66493-39-8

IUPAC Name: (1S)-1-{[1,1'-biphenyl]-4-yl}ethan-1-aminium
Synonyms: (1S)-1-{[1,1'-biphenyl]-4-yl}ethanaminium
Molecular Weight (g/mol): 198.29
Molecular Formula: C14H16N
InChi Key: QVLZRPPCCDKMPO-NSHDSACASA-O
SMILES: C[C@H]([NH3+])C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results