accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 665-46-3

IUPAC Name: tetraethylazanium fluoride
Synonyms: tetraethylammonium fluoride
Molecular Weight (g/mol): 149.25
Molecular Formula: C8H20FN
InChi Key: QSUJAUYJBJRLKV-UHFFFAOYSA-M
SMILES: [F-].CC[N+](CC)(CC)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results