accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6655-27-2

H3CNNHSOONOOCH3

CAS RN 6655-27-2

IUPAC Name: 2-nitro-N'-(propan-2-ylidene)benzene-1-sulfonohydrazide
Synonyms: 2-nitro-N'-(propan-2-ylidene)benzenesulfonohydrazide
Molecular Weight (g/mol): 257.26
Molecular Formula: C9H11N3O4S
InChi Key: SBNYNTYNEJTMQO-UHFFFAOYSA-N
SMILES: CC(C)=NNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results