accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66576-71-4

IUPAC Name: propan-2-yl 2-methylbutanoate
Synonyms: isopropyl 2-methylbutanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: DIRDKDDFAMNBNY-UHFFFAOYNA-N
SMILES: CCC(C)C(=O)OC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results