accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66582-85-2

IUPAC Name: 4-ethyl-1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-imidazol-3-ium-3-olate
Synonyms: 5-ethyl-3-hydroxy-2,2,4,4-tetramethylimidazol-1-ium-1-olate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: GCQNOXDEKPIVOY-UHFFFAOYSA-N
SMILES: CCC1=[N+]([O-])C(C)(C)N(O)C1(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)