accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66584-32-5

IUPAC Name: 3-bromo-N-methylaniline
Synonyms: 3-bromo-N-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: HKOSFZXROYRVJT-UHFFFAOYSA-N
SMILES: CNC1=CC=CC(Br)=C1
Boiling Point 
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results