accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66605-58-1

OOHOOHN(S)CH3H3CCH3

CAS RN 66605-58-1

IUPAC Name: tert-butyl N-[(2S)-1-[4-(benzyloxy)phenyl]-3-hydroxypropan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-[4-(benzyloxy)phenyl]-3-hydroxypropan-2-yl]carbamate
Molecular Weight (g/mol): 357.45
Molecular Formula: C21H27NO4
InChi Key: FOMAWYQEBPYJJU-SFHVURJKSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=C(OCC2=CC=CC=C2)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)