accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 666174-80-7

IUPAC Name: 2,5-dihydroxybenzoic acid; butan-1-amine
Synonyms: 2,5-dihydroxybenzoic acid; butylamine
Molecular Weight (g/mol): 227.26
Molecular Formula: C11H17NO4
InChi Key: OGLUKKYDTRVMHP-UHFFFAOYSA-N
SMILES: CCCCN.OC(=O)C1=C(O)C=CC(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results