accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 666747-06-4

IUPAC Name: (2,4-dibromophenyl)methanol
Synonyms: (2,4-dibromophenyl)methanol
Molecular Weight (g/mol): 265.93
Molecular Formula: C7H6Br2O
InChi Key: VXEVXBMLHQPKGA-UHFFFAOYSA-N
SMILES: OCC1=C(Br)C=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)