accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66710-66-5

IUPAC Name: 2-(ethenesulfonyl)-N-{2-[2-(ethenesulfonyl)acetamido]ethyl}acetamide
Synonyms: 2-(ethenesulfonyl)-N-{2-[2-(ethenesulfonyl)acetamido]ethyl}acetamide
Molecular Weight (g/mol): 324.37
Molecular Formula: C10H16N2O6S2
InChi Key: QWZOJDWOQYTACD-UHFFFAOYSA-N
SMILES: C=CS(=O)(=O)CC(=O)NCCNC(=O)CS(=O)(=O)C=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results