accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66735-20-4

IUPAC Name: propan-2-amine hydroiodide
Synonyms: isopropylamine hydroiodide
Molecular Weight (g/mol): 187.02
Molecular Formula: C3H10IN
InChi Key: VMLAEGAAHIIWJX-UHFFFAOYSA-N
SMILES: I.CC(C)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results