accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOO

CAS RN 66742-57-2

IUPAC Name: 4-[(3-fluorophenyl)methoxy]benzaldehyde
Synonyms: 4-[(3-fluorophenyl)methoxy]benzaldehyde
Molecular Weight (g/mol): 230.24
Molecular Formula: C14H11FO2
InChi Key: DNKSIIHRKWTIRH-UHFFFAOYSA-N
SMILES: FC1=CC=CC(COC2=CC=C(C=O)C=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results