accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66748-43-4

IUPAC Name: 1,3-bis[4-(1-{[(diaminomethylidene)amino]imino}ethyl)phenyl]urea; bis(4-methylbenzene-1-sulfonic acid)
Synonyms: 1,3-bis[4-(1-{[(diaminomethylidene)amino]imino}ethyl)phenyl]urea; bis(para-toluene sulfonate)
Molecular Weight (g/mol): 752.87
Molecular Formula: C33H40N10O7S2
InChi Key: DIQGNVANOSOABS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC1=CC=C(C=C1)S(O)(=O)=O.CC(=NN=C(N)N)C1=CC=C(NC(=O)NC2=CC=C(C=C2)C(C)=NN=C(N)N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results