accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONOO

CAS RN 66750-10-5

IUPAC Name: 13-phenyl-1,4,7,10-tetraoxa-13-azacyclopentadecane
Synonyms: 13-phenyl-1,4,7,10-tetraoxa-13-azacyclopentadecane
Molecular Weight (g/mol): 295.38
Molecular Formula: C16H25NO4
InChi Key: SGDQOAKAHLFKBV-UHFFFAOYSA-N
SMILES: C1COCCOCCN(CCOCCO1)C1=CC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results