accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66778-17-4

IUPAC Name: bis(7-hydroxy-6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one) trihydrate
Synonyms: bis(esculin) trihydrate
Molecular Weight (g/mol): 734.61
Molecular Formula: C30H38O21
InChi Key: LGMHQDJZWHRZPO-UHFFFAOYNA-N
SMILES: O.O.O.OCC1OC(OC2=C(O)C=C3OC(=O)C=CC3=C2)C(O)C(O)C1O.OCC1OC(OC2=C(O)C=C3OC(=O)C=CC3=C2)C(O)C(O)C1O
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)