accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66793-76-8

IUPAC Name: ethyl(heptyl)amine
Synonyms: ethyl(heptyl)amine
Molecular Weight (g/mol): 143.27
Molecular Formula: C9H21N
InChi Key: IUZZLNVABCISOI-UHFFFAOYSA-N
SMILES: CCCCCCCNCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)