accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 668467-91-2

IUPAC Name: {5-chloro-1-[(2,5-dichlorophenyl)methyl]-2-oxo-2,3-dihydro-1H-indol-3-ylidene}amino acetate
Synonyms: {5-chloro-1-[(2,5-dichlorophenyl)methyl]-2-oxoindol-3-ylidene}amino acetate
Molecular Weight (g/mol): 397.64
Molecular Formula: C17H11Cl3N2O3
InChi Key: OPQRFPHLZZPCCH-UHFFFAOYSA-N
SMILES: CC(=O)ON=C1C(=O)N(CC2=C(Cl)C=CC(Cl)=C2)C2=C1C=C(Cl)C=C2
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results