accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCl(S)(S)OHCH3NHOH3CH3C

CAS RN 66866-68-0

IUPAC Name: (2S,3S)-3-methyl-2-(methylamino)pentanoic acid hydrochloride
Synonyms: N-methylisoleucine hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: MMBGZHHBWVCZFK-YMGXCRELNA-N
SMILES: Cl.CC[C@H](C)[C@H](NC)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)