accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 669050-69-5

IUPAC Name: 1H-indazole-6-carbaldehyde
Synonyms: 1H-indazole-6-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: JTWYTTXTJFDYAG-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2C=NNC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results