accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 669050-72-0

IUPAC Name: 3-(piperidin-1-yl)benzaldehyde
Synonyms: 3-(piperidin-1-yl)benzaldehyde
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: FXXQUTSXXILOMY-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CC=C1)N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results