accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClNH2N

CAS RN 66909-38-4

IUPAC Name: 6-chloro-4-methylpyridin-3-amine
Synonyms: 6-chloro-4-methylpyridin-3-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: PLSMBLIGZJWLEJ-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC=C1N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results