accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OBrCH3CH3NCH3(S)(R)OOOH

CAS RN 66985-17-9

IUPAC Name: (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium hydrate bromide
Synonyms: (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-isopropyl-8-methyl-8-azabicyclo[3.2.1]octan-8-ium hydrate bromide
Molecular Weight (g/mol): 430.38
Molecular Formula: C20H32BrNO4
InChi Key: KEWHKYJURDBRMN-XFQAGIBXSA-M
SMILES: O.[Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)