CAS RN 67-66-3
Chloroform, ACS, 99.8+%
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 SMILES: ClC(Cl)Cl
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 SMILES: ClC(Cl)Cl
Chloroform (Ethanol as Preservative/Certified ACS), Fisher Chemical™
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl
Chloroform, 99.8% min., ACS, MilliporeSigma™
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl
Chloroform, 99.9% min., OmniSolv™, MilliporeSigma™
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl
Thermo Scientific Chemicals Chloroform, Molecular Biology Reagent
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 SMILES: ClC(Cl)Cl
Chloroform, HPLC Grade, 99.5+% min
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 SMILES: ClC(Cl)Cl
Chloroform (Approx. 0.75% Ethanol as Preservative/HPLC), Fisher Chemical™
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl
Chloroform/isoamyl alcohol 24:1(v/v), for molecular biology, DNAse, RNAse and Protease free
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Chloroform Molecular Biology MP Biomedicals
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 SMILES: ClC(Cl)Cl
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 SMILES: ClC(Cl)Cl
Chloroform, 99+%, extra pure, stabilized with ethanol
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 IUPAC Name: chloroform SMILES: ClC(Cl)Cl
Chloroform, HPLC Grade, 99.5+% min, stab. with amylene
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl
Chloroform (Ethanol as Preservative, Spectranalyzed™), Fisher Chemical™
CAS: 67-66-3 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio PubChem CID: 6212 ChEBI: CHEBI:35255 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl