accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3OO

CAS RN 67-92-5

IUPAC Name: 2-(diethylamino)ethyl [1,1'-bi(cyclohexane)]-1-carboxylate hydrochloride
Synonyms: dicyclomine hydrochloride
Molecular Weight (g/mol): 345.95
Molecular Formula: C19H36ClNO2
InChi Key: GUBNMFJOJGDCEL-UHFFFAOYSA-N
SMILES: Cl.CCN(CC)CCOC(=O)C1(CCCCC1)C1CCCCC1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results