accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONN

CAS RN 6705-33-5

IUPAC Name: (pyrazin-2-yl)methanol
Synonyms: pyrazin-2-ylmethanol
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: LFCWHDGQCWJKCG-UHFFFAOYSA-N
SMILES: OCC1=CN=CC=N1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)