accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClH2NNOO

CAS RN 67073-39-6

IUPAC Name: 4-chloro-5-nitrobenzene-1,2-diamine
Synonyms: 4-chloro-5-nitrobenzene-1,2-diamine
Molecular Weight (g/mol): 187.58
Molecular Formula: C6H6ClN3O2
InChi Key: LOQLMWFVXRZASN-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)