accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67099-34-7

IUPAC Name: N,N-dibutyl-4-chlorobutanamide
Synonyms: N,N-dibutyl-4-chlorobutanamide
Molecular Weight (g/mol): 233.78
Molecular Formula: C12H24ClNO
InChi Key: UJIBJKONJLJLAU-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results