accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 671-42-1

IUPAC Name: 3-aminoazepan-2-one
Synonyms: α-amino-epsilon-caprolactam 2-aminohexano-6-lactam
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: BOWUOGIPSRVRSJ-UHFFFAOYNA-N
SMILES: NC1CCCCNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results