accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NHNCH3CH3

CAS RN 6711-48-4

IUPAC Name: (3-{[3-(dimethylamino)propyl]amino}propyl)dimethylamine
Synonyms: tetramethyldipropylenetriamine
Molecular Weight (g/mol): 187.33
Molecular Formula: C10H25N3
InChi Key: BXYVQNNEFZOBOZ-UHFFFAOYSA-N
SMILES: CN(C)CCCNCCCN(C)C
Boiling Point 
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)