accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 671789-61-0

IUPAC Name: iridium(3+) 2,6-bis[(di-tert-butylphosphanyl)oxy]benzen-1-ide chloride hydride
Synonyms: iridium(3+) 2,6-bis[(di-tert-butylphosphanyl)oxy]benzen-1-ide chloride hydride
Molecular Weight (g/mol): 626.17
Molecular Formula: C22H40ClIrO2P2
InChi Key: HELSKPCZFUCUCP-UHFFFAOYSA-M
SMILES: [H-].[Cl-].[Ir+3].CC(C)(C)P(OC1=CC=CC(OP(C(C)(C)C)C(C)(C)C)=[C-]1)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results