accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrH3COONHCH3

CAS RN 672-99-1

IUPAC Name: 4-bromo-3,5-dimethylphenyl N-methylcarbamate
Synonyms: 4-bromo-3,5-dimethylphenyl N-methylcarbamate
Molecular Weight (g/mol): 258.12
Molecular Formula: C10H12BrNO2
InChi Key: ZOZFMTULOYRWEL-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC(C)=C(Br)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results