accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNH2NH2O

CAS RN 6720-02-1

IUPAC Name: 2-amino-3-(1H-indol-3-yl)propanamide
Synonyms: tryptophan amide
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: JLSKPBDKNIXMBS-UHFFFAOYNA-N
SMILES: NC(CC1=CNC2=CC=CC=C12)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)