CAS RN 67219-55-0
CAS RN 67219-55-0
IUPAC Name:
N-[1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl]benzamide
Synonyms:
N-[1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl)-2-oxopyrimidin-4-yl]benzamide
Molecular Weight (g/mol):
633.7
Molecular Formula:
C37H35N3O7
InChi Key:
MYSNCIZBPUPZMQ-UHFFFAOYNA-N
SMILES:
COC1=CC=C(C=C1)C(OCC1OC(CC1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
1
–
1
of
1
results
1
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine 99.0+%, TCI America™
CAS: 67219-55-0 Molecular Formula: C37H35N3O7 Molecular Weight (g/mol): 633.701 MDL Number: MFCD00010114 InChI Key: MYSNCIZBPUPZMQ-VOTWKOMSSA-N Synonym: Bz-DMT-dC PubChem CID: 2724490 IUPAC Name: N-[1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide SMILES: COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)OC)OCC4C(CC(O4)N5C=CC(=NC5=O)NC(=O)C6=CC=CC=C6)O