accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6723-30-4

IUPAC Name: O-(oxan-2-yl)hydroxylamine
Synonyms: O-(oxan-2-yl)hydroxylamine
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: NLXXVSKHVGDQAT-UHFFFAOYNA-N
SMILES: NOC1CCCCO1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)