accessibility menu, dialog, popup

UserName

Viewing profile as user:
FePPNiClClPhPhPhPh

CAS RN 67292-34-6

IUPAC Name: Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)
Synonyms:
Molecular Weight (g/mol): 683.98
Molecular Formula: C34H28Cl2FeNiP2
InChi Key:
SMILES: [Fe].Cl[Ni]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)