accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 673-40-5

IUPAC Name: 4-bromobenzene-1-diazonium; tetrafluoroboranuide
Synonyms: 4-bromobenzenediazonium tetrafluoroborate
Molecular Weight (g/mol): 270.82
Molecular Formula: C6H4BBrF4N2
InChi Key: FXTCQUCYDJZGGU-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)