CAS RN 6731-36-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane, 75% solution in aromatic free mineral spirit
CAS: 6731-36-8 | C2H4Cl2 | 98.954 g/mol