accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6734-33-4

CH3OOO(S)O(R)OH(S)OH(R)HO

CAS RN 6734-33-4

IUPAC Name: 4-methyl-7-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}-2H-chromen-2-one
Synonyms: 4-methyl-7-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}chromen-2-one
Molecular Weight (g/mol): 308.29
Molecular Formula: C15H16O7
InChi Key: JWIYLOHVJDJZOQ-KAOXEZKKSA-N
SMILES: CC1=CC(=O)OC2=CC(O[C@@H]3OC[C@@H](O)[C@H](O)[C@H]3O)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results