accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CBrBrNCH3CH3

CAS RN 673474-73-2

IUPAC Name: (3-{2,7-dibromo-9-[3-(dimethylamino)propyl]-9H-fluoren-9-yl}propyl)dimethylamine
Synonyms: (3-{2,7-dibromo-9-[3-(dimethylamino)propyl]fluoren-9-yl}propyl)dimethylamine
Molecular Weight (g/mol): 494.32
Molecular Formula: C23H30Br2N2
InChi Key: RJIWYGUYDXBQCJ-UHFFFAOYSA-N
SMILES: CN(C)CCCC1(CCCN(C)C)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results